missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Antibody (CL13628) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24566AF532
This item is not returnable.
View return policy
Description
YAP1 Monoclonal antibody specifically detects YAP1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| YAP1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
| This antibody was developed using a synthetic peptide derived from P46937, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
| 10413 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL13628 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction