missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylometazoline EP Impurity A (as Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Xylometazoline EP Impurity A (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | N-(2-Aminoethyl)-2-[4-(1,1-dimethylethyl)-2,6-dimethylphenyl]acetamide Hydrochloride |
| CAS | 81201-80-1 |
| Analyte or Component Names | N-(2-Aminoethyl)-2-[4-(1,1-dimethylethyl)-2,6-dimethylphenyl]acetamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H26 N2 O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Xylometazoline Hydrochloride Imp. A (EP), Xylometazoline Imp. A (EP), N-(2-Aminoethyl)-2-[4-(1,1-dimethylethyl)-2,6-dimethylphenyl]acetamide Hydrochloride, Xylometazoline Hydrochloride Imp. A (EP) as Hydrochloride, Xylometazoline Related Compound A as Hydrochloride, Xylometazoline Hydrochloride Impurity A as Hydrochloride, Xylometazoline Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C16H26N2O.ClH/c1-11-8-13(16(3,4)5)9-12(2)14(11)10-15(19)18-7-6-17;/h8-9H,6-7,10,17H2,1-5H3,(H,18,19);1H |
| SMILES | Cl.Cc1cc(cc(C)c1CC(=O)NCCN)C(C)(C)C |
| IUPAC Name | N-(2-aminoethyl)-2-(4-tert-butyl-2,6-dimethylphenyl)acetamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction