missing translation for 'onlineSavingsMsg'
Learn More

XPA Antibody (2A4), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18734032 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18734032 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18734032 Supplier Novus Biologicals Supplier No. NBP250600AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

XPA Monoclonal antibody specifically detects XPA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPA
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 2A4
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias DNA repair protein complementing XP-A cells, excision repair-controlling, Xeroderma pigmentosum group A-complementing protein, xeroderma pigmentosum, complementation group A, XPACXP1
Host Species Mouse
Immunogen His-tagged recombinant human XPA protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 7507
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.