Learn More
SNV Comines™ Xavier™ Cotton Laboratory Coats
Description
- White in color
- With snap buttons
- Collared with lapels
Specifications
Specifications
| Product Type | Xavier Lab Coat |
| Size | 46 |
| Gender | Men's |
| Material | Carbon |
| Color | White |
| Closure Type | Snaps |
| No. of Pockets | 4 |
| Size (Metric) Chest | 100 cm |
| Includes | 2 Flat pockets, 1 breast pocket, 1 inside pocket, opening angle |
| Construction | 100% Cotton |