missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XAGE5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 624.00€
Specifications
| Antigen | XAGE5 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18294105
|
Novus Biologicals
NBP2-58687 |
100 μL |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621628
|
Novus Biologicals
NBP2-58687-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XAGE5 Polyclonal specifically detects XAGE5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| XAGE5 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 170627 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GRRYRPRRCLRLAQLVGPMLEPSVPEPQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Cancer/Testis Antigen 12.5, Cancer/Testis Antigen Family 12, Member 5, CT12.5, G Antigen Family D Member 5, G Antigen, Family D, 5, GAGED5, Protein XAGE-5, X Antigen Family Member 5, XAGE-5 | |
| XAGE5 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title