missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WSB2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94083-0.02ml
This item is not returnable.
View return policy
Description
WSB2 Polyclonal antibody specifically detects WSB2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| WSB2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| CS box-containing WD protein, MGC10210, SBA2, WD repeat and SOCS box containing 2, WD repeat and SOCS box containing protein 2, WD repeat and SOCS box-containing 2, WD repeat and SOCS box-containing protein 2, WSB-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human WSB2 (NP_061109.1). MEAGEEPLLLAELKPGRPHQFDWKSSCETWSVAFSPDGSWFAWSQGHCIVKLIPWPLEEQFIPKGFEAKSRSSKNETKGRGSPKEKTLDCGQIVWGLAFSPWPSPPSRKLWARHHPQVPDVSCLVLATGLNDGQIKIWEVQTGLLLLNLSGHQDVVRDLSFTPSGSLILVSASRDKTLRIWDLNKHGKQIQVLSGHLQWV | |
| 0.02 mL | |
| Signal Transduction | |
| 55884 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction