missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT10A Monoclonal Antibody (342811)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561312
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is strongly expressed in the cell lines of promyelocytic leukemia and Burkitt's lymphoma. In addition, it and another family member, the WNT6 gene, are strongly coexpressed in colorectal cancer cell lines. The gene overexpression may play key roles in carcinogenesis through activation of the WNT-beta-catenin-TCF signaling pathway. This gene and the WNT6 gene are clustered in the chromosome 2q35 region.
Specifications
| WNT10A | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| P70701, Q9GZT5 | |
| WNT10A | |
| E. coli-derived recombinant mouse Wnt-10a, Leu55-Glu95 and Gln278-Arg360. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 342811 | |
| Unconjugated | |
| WNT10A | |
| OODD; Protein Wnt-10a; SSPS; STHAG4; wingless related MMTV integration site 10a; wingless-type MMTV integration site family member 10A; wingless-type MMTV integration site family, member 10A; Wnt family member 10A; Wnt10a | |
| Rat | |
| Protein A/G | |
| RUO | |
| 22409, 80326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction