Learn More
Wnt-5a Antibody (S08-4A3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20061-100ul
 View more versions of this product
Product Code. 18603084
  • 645.00€ / 100µL
Description
Wnt-5a Monoclonal antibody specifically detects Wnt-5a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), RNA Inhibition, Dot Blot, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wnt-5a
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
S08-4A3
Unconjugated
hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
A synthetic peptide of human Wnt-5a (Uniprot # P41221)
100 μg
Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
7474
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.