Learn More
DWK Life Sciences Wheaton™ LDPE Dropping Bottles
Product Code. 12466171 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
3 mL
6 mL
7 mL
10 mL
15 mL
30 mL
60 mL
Closure Size:
8-425 mm
13-425 mm
15-415 mm
20-410 mm
Color:
Natural
White
Quantity:
100/Cs.
1000/Cs.
Unit Size:
Pack of 100
Pack of 1000
Product Code. 12466171
Brand: DWK Life Sciences W242822
Only null left
Add to basket
Add to basket
Excellent chemical resistance
  • Available in a variety of sizes
  • Packaged in cases of 100, 1000 or bulk
Order Info
Dropper tips and caps must be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 6 mL, 0.2 oz.
Capacity (Metric) 6 mL
Capacity (English) 0.2 oz.
Color Natural
Diameter (English) 0.66 in.
Material LDPE
No. per Pack 100/Cs.
Quantity 100/Cs.
Closure Included No
Height (Metric) 42 mm
Height (English) 1.65 in.
Closure Material Cellulose Acetate Proprionate
Closure Size 13-425 mm
Diameter (Metric) 17 mm
Autoclavable Not Autoclavable