Learn More
DWK Life Sciences Wheaton™ Amber Glass Dropping Bottle
Product Code. 12329719 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
Color:
Amber
Clear
Unit Size:
Pack of 6
Product Code. 12329719
Brand: DWK Life Sciences W211755
Only null left
Add to basket
Add to basket
Delivers stains or indicator solutions drop by drop
  • Complete with a glass stopper
  • A partial turn of the ground glass stopper seals the bottle
  • Manufactured from soda-lime glass that conforms to USP Type III requirements
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Capacity 3.38 oz., 100 mL
Capacity (Metric) 100 mL
Capacity (English) 3.38 oz.
Color Amber
Diameter (English) 2.2 in.
Includes Pouring spout
Material Glass
Shape Round
Height (Metric) 120 mm
Product Type Dropping Bottle
Height (English) 4.72 in.
Diameter (Metric) 56 mm
Closure Type Stopper