Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Featuring sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters
94.60€ - 1140.00€
Specifications
Material | Polycarbonate |
---|---|
For Use With (Application) | Epifluorescence Microscopy, Environmental Analysis, Cell Biology, EPA Testing, Fuel Testing, Bioassays, Parasitology, Air Analysis, Water Microbiology |
Type | Membrane Circle |
Product Code | Brand | Dimensions (L x W) | Pore Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Pore Size | Quantity | Price | Quantity & Availability | |||
17149880
|
Cytiva
10417050 |
25 Pk. |
545.00€
Pack of 25 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
17199880
|
Cytiva
10417551 |
209.55 x 292.1 mm | 0.5 μm | 25 Pk. |
1140.00€
Pack of 25 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||
17189880
|
Cytiva
10417451 |
5 μm | 100 Pk. |
94.60€
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
17806797
|
Cytiva
10417450 |
25 x 80 mm | 5 μm | 100 Pk. |
128.00€
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
17159880
|
Cytiva
10417051 |
0.2 μm | 25 Pk. |
915.00€
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
17169880
|
Cytiva
10417139 |
293 mm | 0.4 μm | 25 Pk. |
915.00€
Pack of 25 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||
17179880
|
Cytiva
10417450 |
100 Pk. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Low affinity for stains improves visibility under microscope
- Low protein binding minimizes sample loss of proteinaceous samples
- Narrow pore size distribution and high pore density support rapid surface capture of analyte
- Negligible absorption and adsorption of filtrate; nonhygroscopic
- No particle shedding enables clean filtrate
- Biologically inert
Specifications
Polycarbonate | |
Epifluorescence Microscopy, Environmental Analysis, Cell Biology, EPA Testing, Fuel Testing, Bioassays, Parasitology, Air Analysis, Water Microbiology | |
Membrane Circle |