Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Featuring sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters
Brand:  Cytiva 10417051
Additional Details : Weight : 0.10700kg
 View more versions of this product
Product Code. 17159880
  • 915.00€ / Pack of 100
Estimated Shipment: 12-06-2024
Description
  • Low affinity for stains improves visibility under microscope
  • Low protein binding minimizes sample loss of proteinaceous samples
  • Narrow pore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically inert
Specifications
Specifications
Polycarbonate
0.2 μm
25 Pk.
Membrane Circle