Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Featuring sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters
Brand: Cytiva 10417450
Additional Details : Weight : 0.82400kg
Description
- Low affinity for stains improves visibility under microscope
- Low protein binding minimizes sample loss of proteinaceous samples
- Narrow pore size distribution and high pore density support rapid surface capture of analyte
- Negligible absorption and adsorption of filtrate; nonhygroscopic
- No particle shedding enables clean filtrate
- Biologically inert
Specifications
Polycarbonate | |
100 Pk. |