missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus Envelope Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41056-0.025mg
This item is not returnable.
View return policy
Description
West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISA.
Specifications
| West Nile Virus Envelope | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide corresponding to 16 amino acids at the N terminus of the West Nile virus envelope protein. The immunogen is located within amino acids 350 - 400 of West Nile Virus Envelope Protein. Amino Acid Squence: CPTMGEAHNEKRADPA | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction