missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDHD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
369.00€ - 500.00€
Specifications
| Antigen | WDHD1 |
|---|---|
| Dilution | Western Blot 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452671
|
Novus Biologicals
NBP1-89091-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726783
|
Novus Biologicals
NBP1-89091 |
500.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
WDHD1 Polyclonal specifically detects WDHD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| WDHD1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| O75717 | |
| 11169 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QTLNIVTWSPCGQYLAAGSINGLIIVWNVETKDCMERVKHEKGYAICGLAWHPTCGRISYTDAEGNLGLLENVCDPSGKTSSSKVSSRVEKDYNDLFDGDDMSNAGDFLNDNAVE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 126 kDa |
| Western Blot 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Acidic nucleoplasmic DNA-binding protein 1, AND1, AND-1, CHTF4, CTF4, CTF4, chromosome transmission fidelity factor 4 homolog, WD repeat and HMG-box DNA binding protein 1, WD repeat and HMG-box DNA-binding protein 1 | |
| WDHD1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human WDHD1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title