missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDHD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93674-0.02ml
This item is not returnable.
View return policy
Description
WDHD1 Polyclonal antibody specifically detects WDHD1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| WDHD1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Acidic nucleoplasmic DNA-binding protein 1, AND1, AND-1, CHTF4, CTF4, CTF4, chromosome transmission fidelity factor 4 homolog, WD repeat and HMG-box DNA binding protein 1, WD repeat and HMG-box DNA-binding protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 830-1129 of human WDHD1 (NP_009017.1). EEEEEEDFRKKLNAGYSNTATEWSQPRFRNQVEEDAEDSGEADDEEKPEIHKPGQNSFSKSTNSSDVSAKSGAVTFSSQGRVNPFKVSASSKEPAMSMNSARSTNILDNMGKSSKKSTALSRTTNNEKSPIIKPLIPKPKPKQASAASYFQKRNSQTNKTEEVKEENLKNVLSETPAICPPQNTENQRPKTGFQMWLEENRSNILSDNPDFSDEADIIKEGMIRFRVLSTEERKVWANKAKGETASEGTEAKKRKRVVDESDETENQEEKAKENLNLSKKQKPLDFSTNQKLSAFAFKQE | |
| 0.02 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 11169 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction