missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Washington Residual Solvents Mixture 709 290-5000 μg/mL in N,N-Dimethylacetamide, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Acetone∣Cyclohexane∣Dichloromethane (Methylenechloride)∣Ethyl acetate∣Isopropyl alcohol∣Methanol∣n-Butane∣n-Heptane∣n-Hexane∣n-Pentane∣N-Propane∣Toluene∣Xylene (all isomers) |
| CAS | 67-64-1, 110-82-7, 75-09-2, 141-78-6, 67-63-0, 67-56-1 |
| Analyte or Component Names | Acetone∣Cyclohexane∣Dichloromethane (Methylenechloride)∣Ethyl acetate∣Isopropyl alcohol∣Methanol∣n-Butane∣n-Heptane∣n-Hexane∣n-Pentane∣N-Propane∣Toluene∣Xylene (all isomers) |
| Shipping Condition | +4°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | N,N-Dimethylacetamide |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | n-Hexane 290 μg/mL∣Dichloromethane (Methylenechloride) 600 μg/mL∣Toluene 890 μg/mL∣Xylene (all isomers) 2170 μg/mL∣Methanol 3000 μg/mL∣Cyclohexane 3880 μg/mL∣Acetone 5000 μg/mL∣Ethyl acetate 5000 μg/mL∣Isopropyl alcohol 5000 μg/mL∣n-Butane 5000 μg/mL∣n-Heptane 5000 μg/mL∣n-Pentane 5000 μg/mL∣N-Propane 5000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction