Learn More
Kartell™ Wash Bottles
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Pack of 10
Description
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| For Use With (Application) | Suitable for foodstuff |
| Material | Polyethylene |
| Quantity | 10 |
| Height (Metric) | 140 mm |
| Diameter (Metric) | 60 mm |