missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VZV gH/gL Antibody [DyLight 755] - (Oka vaccine), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43771IR
This item is not returnable.
View return policy
Description
VZV gH/gL Polyclonal antibody specifically detects VZV gH/gL in Virus samples. It is validated for ELISA,Western Blot
Specifications
| VZV gH/gL | |
| Polyclonal | |
| Western Blot, ELISA | |
| envelope glycoprotein H, Envelope glycoprotein L | |
| Produced in rabbits immunized with purified, recombinant Varicella Zoster Virus gH & gL Heterodimer (Oka vaccine) (Uniprot#: Q775J3 & Q9J3N1) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| DyLight 755 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction