missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558136
This item is not returnable.
View return policy
Description
Immunogen sequence: VNPDVTQVGL VVYGSQVQTA FGLDTKPTRA AMLRAISQAP YLGGVGSAGT ALLHIYDKVM TVQRGARPGV PKAVVVLTGG RGAEDAAVPA QKLRNNGISV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
VWA2 Polyclonal antibody specifically detects VWA2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| VWA2 | |
| Polyclonal | |
| Unconjugated | |
| VWA2 | |
| a domain-containing protein similar to matrilin and collagen; AMACO; CCSP-2; colon cancer diagnostic marker; colon cancer secreted protein 2; NET42; von Willebrand factor A domain containing 2; von Willebrand factor A domain-containing protein 2; VWA2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 340706 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5GFL6 | |
| VWA2 | |
| Recombinant protein corresponding to Human VWA2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction