missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSTM2A Monoclonal Antibody (1024735)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562079
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Plays a role in the regulation of the early stage of white and brown preadipocyte cell differentiation. Promotes adipogenic commitment of preadipocytes by increasing gene expression of the transcription factor PPARG in a BMP4-dependent signaling pathway.
Specifications
| VSTM2A | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8TAG5 | |
| VSTM2A | |
| Human embryonic kidney cell HEK293-derived human VSTM2A protein, Ser25-Phe244. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry | |
| 1024735 | |
| Unconjugated | |
| VSTM2A | |
| hypothetical protein MGC33530; V-set and transmembrane domain containing 2; V-set and transmembrane domain containing 2A; V-set and transmembrane domain-containing protein 2A; Vstm2; VSTM2A | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 222008 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction