missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS25 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | VPS25 |
|---|---|
| Clone | 3B4C3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233173
|
Proteintech
68197-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236155
|
Proteintech
68197-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The VPS25 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VPS25. This antibody recognizes human, rabbit antigen. The VPS25 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| VPS25 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 84313 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 3B4C3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Rabbit | |
| DERP9; EAP20; FAP20; MGC10540 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title