Learn More
VPREB3 Antibody (OTI1A3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45027-100ul
This item is not returnable.
View return policy
Description
VPREB3 Monoclonal antibody specifically detects VPREB3 in Human samples. It is validated for Western Blot
Specifications
| VPREB3 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1A3 | |
| Western Blot 1:2000 | |
| N27C7-2, pre-B lymphocyte 3,8HS20, pre-B lymphocyte gene 3, pre-B lymphocyte protein 3, Protein VPreB3 | |
| Human recombinant protein fragment corresponding to amino acids 48-123 of human VPREB3 (NP_037510) produced in E.coli. | |
| 100 μL | |
| Adaptive Immunity, Immunology | |
| 29802 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |