missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vortioxetine, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Vortioxetine |
| CAS | 508233-74-7 |
| Analyte or Component Names | Vortioxetine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine, Lu AA 21004, Vortioxetine |
| InChI Formula | InChI=1S/C18H22N2S/c1-14-7-8-17(15(2)13-14)21-18-6-4-3-5-16(18)20-11-9-19-10-12-20/h3-8,13,19H,9-12H2,1-2H3 |
| Solution Type | Neat |
| SMILES | Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction