missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Voriconazole Anti-Fungal Agent, Solid, 349.31 Da, 50 mg
Description
- Molecular Weight: 349.31 Da
- Purity: >99%
- Form: Solid
- Description: Potent and broad-spectrum anti-fungal agent that inhibits CYP450-dependent 14-α sterol demethylase, interfering with ergosterol biosynthesis and inhibiting fungal growth.
- Storage: Store at +4°C.
- Shipping: Ambient - Can Ship with Ice
- Composition: 100% Voriconazole
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Alternate Part Number | UK-109, 496 |
| Buffer | 100% Voriconazole |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | 14-α Sterol Demethylase |
| Formulation | 100% Voriconazole |
| Quantity | 50 mg |
| For Use With (Application) | Fungal growth inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?