missing translation for 'onlineSavingsMsg'
Learn More

Von Willebrand Factor Rabbit anti-Human, Clone: VWF/1859R, Novus Biologicals™

Rabbit Monoclonal Antibody

528.00€

Specifications

Antigen Von Willebrand Factor
Clone VWF/1859R
Concentration 0.2 mg/mL
Dilution Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18674719
View Documents
Novus Biologicals
NBP3-07217-100ug
100 μg
528.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

Von Willebrand Factor
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Recombinant fragment of human Von Willebrand Factor protein (aa1815-1939) (exact sequence is proprietary)
Primary
Store at 4°C.
VWF/1859R
Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
7450
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.