missing translation for 'onlineSavingsMsg'
Learn More

VN1R3 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16803155 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16803155 20 μL 20µL
16893145 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16803155 Supplier Proteintech Supplier No. 246591AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Putative pheromone receptor. There seems to be a deletion in the gene coding for this protein in about 98% of the population. The gorilla and orangutan orthologous proteins do not exist, their genes are pseudogenes.

Specifications

Antigen VN1R3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene VN1R3
Gene Accession No. Q9BXE9
Gene Alias FKSG46, V1RL3, V1RL3p, VN1R3
Gene Symbols VN1R3
Host Species Rabbit
Immunogen VN1R3 Fusion Protein Ag20382
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317702
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.