missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VMAT2 Antibody (CL13157), Novus Biologicals™
Mouse Monoclonal Antibody
429.00€ - 611.00€
Specifications
| Antigen | VMAT2 |
|---|---|
| Clone | CL13157 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600035
|
Novus Biologicals
NBP3-18556-100ul |
100 μg |
611.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653435
|
Novus Biologicals
NBP3-18556-25ul |
25 μg |
429.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VMAT2 Monoclonal antibody specifically detects VMAT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VMAT2 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2 | |
| This antibody was developed against a sythetic peptide for Q05940. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL13157 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| PBS, pH 7.2, 40% glycerol | |
| 6571 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title