missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLDLR Rabbit anti-Human, Rat, Clone: VLDLR/2896R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07198-100ug
This item is not returnable.
View return policy
Description
VLDLR Monoclonal specifically detects VLDLR in Human, Rat samples. It is validated for ELISA.
Specifications
| VLDLR | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA | |
| CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH | |
| Recombinant human VLDLR fragment from c-terminal | |
| 100 μg | |
| Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 7436 | |
| Store at 4°C. | |
| IgG |
| ELISA | |
| VLDLR/2896R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction