missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral wnvNS3 Protease Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF2907-SP
This item is not returnable.
View return policy
Description
wnvNS3 Protease Polyclonal specifically detects wnvNS3 Protease in Viral samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| wnvNS3 Protease | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| flavivirus polyprotein gene | |
| E. coli-derived recombinant wnvNS3 Protease, expressed as a fusion protein with cofactor NS2L Gly1506-Leu1689 Accession # ABD85079 | |
| 25 μg | |
| Primary | |
| Detects West Nile Virus NS3 Protease in direct ELISAs and Western blots. | |
| Virus | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| ABD85079 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 912267 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction