missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIPAR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94018-0.1ml
This item is not returnable.
View return policy
Description
VIPAR Polyclonal antibody specifically detects VIPAR in Mouse, Rat samples. It is validated for Western Blot
Specifications
| VIPAR | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C14orf133FLJ12707, hSPE-39, Protein spe-39 homolog, SPE-39, SPE39chromosome 14 open reading frame 133, VPS16B, VPS33B interacting protein, apical-basolateral polarity regulator, VPS33B-interacting protein, VPS33B-interacting protein involved in polarity and apical protein restriction | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human VIPAS39 (NP_071350.2). MNRTKGDEEEYWNSSKFKAFTFDDEDDELSQLKESKRAVNSLRDFVDDDDDDDLERVSWSGEPVGSISWSIRETAGNSGSTHEGREQLKSRNSFSSYAQLPKPTSTYSLSSFFRGRTRPGSFQSLSDALSDTPAKSYAPELGRPKGEYRDYSNDWSPSDTVRRLRKGKVCSLERFRSLQDKLQLLEEAVSMHDGNVITAVLIFLKRTLSKEILFRELEVR | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 63894 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction