missing translation for 'onlineSavingsMsg'
Learn More

Vincristine sulfate, 97+%, Thermo Scientific™

An anti-tumor alkaloid used in cancer research

185.00€ - 356.00€

Chemical Identifiers

CAS 2068-78-2
Molecular Formula C46H58N4O14S
Molecular Weight (g/mol) 923.044
MDL Number MFCD00084729
InChI Key AQTQHPDCURKLKT-DJOZZVTQSA-N
PubChem CID 126963557
SMILES CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15483589
SDS View Documents
Thermo Scientific Alfa Aesar
J60907.MA
10mg
185.00€
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15493589
SDS View Documents
Thermo Scientific Alfa Aesar
J60907.MB
25mg
356.00€
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in DMSO at 60mg/ml. Soluble in water at 25mg/ml with warming. Very poorly soluble in ethanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents, heavy metal salts.
Specifications

Chemical Identifiers

2068-78-2
923.044
AQTQHPDCURKLKT-DJOZZVTQSA-N
CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
C46H58N4O14S
MFCD00084729
126963557

Specifications

2068-78-2
C46H58N4O14S
UN2811
273°C to 281°C
Soluble in DMSO at 60mg/ml; Soluble in water at 25mg/ml with warming; Very poorly soluble in ethanol
CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
126963557
≥98%
White
MFCD00084729
3924631
14,9986
AQTQHPDCURKLKT-DJOZZVTQSA-N
923.044
923.04
Vincristine sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H341
May damage fertility or the unborn child.
Suspected of causing genetic defects.

P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H301-H341-H361fd

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 218-190-0

missing translation for 'rtecsNumber' : OH6340000

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

Product Identifier
  • Vincristine sulfate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H341-Suspected of causing genetic defects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Vincristine sulfate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.