Learn More
Vincristine sulfate, 97+%, Thermo Scientific™
An anti-tumor alkaloid used in cancer research
Brand: Thermo Scientific Alfa Aesar J60907.MA
| Quantity | 10mg |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in DMSO at 60mg/ml. Soluble in water at 25mg/ml with warming. Very poorly soluble in ethanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents, heavy metal salts.
Chemical Identifiers
| 2068-78-2 | |
| 923.044 | |
| AQTQHPDCURKLKT-DJOZZVTQSA-N | |
| CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O |
| C46H58N4O14S | |
| MFCD00084729 | |
| 126963557 |
Specifications
| 2068-78-2 | |
| 10mg | |
| MFCD00084729 | |
| 3924631 | |
| 14,9986 | |
| AQTQHPDCURKLKT-DJOZZVTQSA-N | |
| 923.044 | |
| 923.04 | |
| Vincristine sulfate |
| White | |
| C46H58N4O14S | |
| UN2811 | |
| 273°C to 281°C | |
| Soluble in DMSO at 60mg/ml; Soluble in water at 25mg/ml with warming; Very poorly soluble in ethanol | |
| CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O | |
| 126963557 | |
| ≥98% |
Safety and Handling
- Vincristine sulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H341-Suspected of causing genetic defects.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Vincristine sulfate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.