missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Vimentin Peptide, Synthetic, Human, >90% Purity, 1 mg
Description
- Protein Name: Vimentin
- Type: Synthetic blocking peptide
- Purity: >90%
- Tested Applications: BL
- Form: Liquid
- Species Reactivity: Human
- Formulation: pH 6.75; 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, and 0.001% Sorbitan monolaurate, ethoxylated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH6.75 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Vimentin |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?