missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Antibody (VM452), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-33060V

Product Code. 18120623

  • 615.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Vimentin Monoclonal specifically detects Vimentin in Human, Porcine, Bovine, Canine, Chicken, Feline, Goat, Mouse (Negative), Rat (Negative) samples. It is validated for Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vimentin
Monoclonal
DyLight 405
FLJ36605, vimentin
Mouse
Protein A or G purified
RUO
Primary
This monoclonal antibody reacts with a 58kDa protein identified as Vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFP however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of Vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-Vimentin. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-Vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain Vimentin, e.g. hepatoma and seminoma. Anti-Vimentin is also useful as a tissue process control reagent.
Store at 4C in the dark.
IgG1 κ
ELISA
VM452
ELISA
VIM
Recombinant full-length human vimentin protein (Uniprot: P08670)
0.1 mL
Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells
7431
Human, Pig, Bovine, Canine, Chicken, Feline, Goat, Mouse (Negative), Rat (Negative)
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only