Learn More
Melford Peptone, Powder
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
Description
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue.
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Low NaCl content suitable for tissue culture.
Total Nitrogen (TN): 15.0-16.0%
Amino Nitrogen (AN): 3.0-3.5%
AN/TN Ratio: 0.21-0.23
Specifications
Specifications
| Quantity | 1 kg |