Learn More
Abnova™ Human SLC6A16 Full-length ORF (AAH34948.1, 1 a.a. - 659 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16158892
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16158892
Brand: Abnova™ H00028968P01.25ug
Used for AP, Array, ELISA, WB-Re
SLC6A16 shows structural characteristics of an Na(+)- and Cl(-)-dependent neurotransmitter transporter, including 12 transmembrane (TM) domains, intracellular N and C termini, and large extracellular loops containing multiple N-glycosylation sites.[supplied by OMIM]
Sequence: MKTEAQPSTSLLANTSWTGTVISDSVPGSQTWEDKGSLTRSATSWTSEAQVSAARVAEAQARTSQPKQISVLEALTASALNQKPTHEKVQMTEKKESEVLLARPFWSSKTEYILAQVGFSMKPSCLWRFAYLWLNSGGCSFAAIYIFMLFLVGVPLLFLEMAAGQSMRQGGMGVWKIIAPWIGGVGYSSFMVCFILGLYFNVVNSWIIFYMSQSFQFPVPWEKCPLTMNSSGFDPECERTTPSIYFWYQQALKASDRIEDGGSPVYSLVLPFFLCWCLVGAFMINGLKSTGKVIYVLVLLPCFIIVGFFIRTLLLEGAKFGLQQLVVAKISDVYNMSVWSLAGGQVLSNTGIGLGSVASLASYMPQSNNCLSDAFLVSVINLLTLLVFTSFNFCVLGFWATVITHRCCERNAEILLKLINLGKLPPDAKPPVNLLYNPTSIYNAWLSGLPQHIKSMVLREVTECNIETQFLKASEGPKFAFLSFVEAMSFLPPSVFWSFIFFLMLLAMGLSSAIGIMQGIITPLQDTFSFFRKHTKLLIVGVFLLMFVCGLFFTRPSGSYFIRLLSDYWIVFPIIVVVVFETMAVSWAYGARRFLADLTILLGHPISPIFGWLWPHLCPVVLLIIFVTMMVHLCMKPITYMSWDSSTVSLPQDPEPLWE
Specifications
Accession Number AAH34948.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 28968
Molecular Weight (g/mol) 99.9kDa
Name SLC6A16 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MKTEAQPSTSLLANTSWTGTVISDSVPGSQTWEDKGSLTRSATSWTSEAQVSAARVAEAQARTSQPKQISVLEALTASALNQKPTHEKVQMTEKKESEVLLARPFWSSKTEYILAQVGFSMKPSCLWRFAYLWLNSGGCSFAAIYIFMLFLVGVPLLFLEMAAGQSMRQGGMGVWKIIAPWIGGVGYSSFMVCFILGLYFNVVNSWIIFYMSQSFQFPVPWEKCPLTMNSSGFDPECERTTPSIYFWYQQALKASDRIEDGGSPVYSLVLPFFLCWCLVGAFMINGLKSTGKVIYVLVLLPCFIIVGFFIRTLLLEGAKFGLQQLVVAKISDVYNMSVWSLAGGQVLSNTGIGLGSVASLASYMPQSNNCLSDAFLVSVINLLTLLVFTSFNFCVLGFWATVITHRCCERNAEILLKLINLGKLPPDAKPPVNLLYNPTSIYNAWLSGLPQHIKSMVLREVTECNIETQFLKASEGPKFAFLSFVEAMSFLPPSVFWSFIFFLMLLAMGLSSAIGIMQGIITPLQDTFSFFRKHTKLLIVGVFLLMFVCGLFFTRPSGSYFIRLLSDYWIVFPIIVVVVFETMAVSWAYGARRFLADLTILLGHPISPIFGWLWPHLCPVVLLIIFVTMMVHLCMKPITYMSWDSSTVSLPQDPEPLWE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NTT5
Common Name SLC6A16
Gene Symbol SLC6A16
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid