Learn More
VPS45 Polyclonal Antibody, Invitrogen™
Product Code. 10240150
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10240150 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10240150 Supplier Invitrogen Supplier No. OSV00016W
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Rabbit Polyclonal Antibody
Glycerol (1:1) may be added for additional stability.
expressed in brain and testis; may play a role in intracellular vesicle mediated transport.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Vps45
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Vps45
Gene Accession No. P97390, O08700
Gene Alias Vps45, VPS45, rvps45, Vps45a
Gene Symbols VPS45
Host Species Rabbit
Immunogen A synthetic peptide from the internal part of rat Vacuolar protein sorting-associated protein 45 (rvps45, Vps45, Vps45a) conjugated to an immunogenic carrier protein was used as the immunogen
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22365, 64516
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG