Learn More
VIAAT/SLC32A1/VGAT Antibody (CL2793) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46628AF532
Product Code. 30291374
  • 737.00€ / 0.10mL
Description
VIAAT/SLC32A1/VGAT Monoclonal antibody specifically detects VIAAT/SLC32A1/VGAT in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VIAAT/SLC32A1/VGAT
Monoclonal
Alexa Fluor 532
bA122O1.1, hVIAAT, solute carrier family 32 (GABA vesicular transporter), member 1, Solute carrier family 32 member 1, Vesicular GABA transporter, vesicular inhibitory amino acid transporter, VGATGABA and glycine transporter, VIAAT
This antibody was developed using a recombinant protein derived from Q9H598, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience, Neurotransmission
140679
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL2793
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat