Learn More
VHL Mouse anti-Human, Clone:, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 556347
This item is not returnable.
View return policy
Description
VHL Monoclonal antibody specifically detects VHL in Human, Mouse samples. It is validated for Immunohistochemistry (Formalin-Antigen Retrieval Required) .
Specifications
| VHL | |
| Monoclonal | |
| Unconjugated | |
| von Hippel-Lindau protein; RCA1; HRCA1 | |
| Human VHL aa. 1-213 Recombinant Protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
| IgG1 κ |
| Immunohistochemistry | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse | |
| Purified |
Product Content Correction
Product Title
For Reseach Use Only.