missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vesicular Stomatitis Indiana Virus Glycoprotein Antibody (110), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-40579C
This item is not returnable.
View return policy
Description
Vesicular Stomatitis Indiana Virus Glycoprotein Monoclonal antibody specifically detects Vesicular Stomatitis Indiana Virus Glycoprotein in Virus samples. It is validated for ELISA
Specifications
| Vesicular Stomatitis Indiana Virus Glycoprotein | |
| Monoclonal | |
| DyLight 650 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA | |
| 110 | |
| 50mM Sodium Borate | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Vesicular Stomatitis Indiana Virus Glycoprotein - (San Juan) (RefSeq#: NP_041715.1; Met1-Ser467). HEK293 used as expression system. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction