missing translation for 'onlineSavingsMsg'
Learn More

Abcam Venlafaxine Hydrochloride, Serotonin-Norepinephrine Reuptake Inhibitor, Solid, 99% Purity, 50 mg

Product Code. 32109870
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109870 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109870 Supplier Abcam Supplier No. ab12071550mg
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent antidepressant and serotonin-norepinephrine reuptake inhibitor with greater than 99% purity.

  • Molecular Weight: 313.87 g/mol
  • Purity: >99%
  • Form: Solid
  • Function: Serotonin-norepinephrine reuptake inhibitor; antidepressant
  • Binding Affinity: Ki values are 82 nM for SERT and 2480 nM for NET; displays 30-fold higher affinity for SERT than NET in vitro
  • Storage: Store at ambient
  • Shipping: Ambient - Can ship with ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Target Molecule SERT, NET
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.