missing translation for 'onlineSavingsMsg'
Learn More

Abcam Venlafaxine Hydrochloride, Serotonin-norepinephrine Reuptake Inhibitor, Solid, 10 mg

Product Code. 32110795
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110795 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110795 Supplier Abcam Supplier No. ab12071510mg
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. Se returpolicy

Potent antidepressant serotonin-norepinephrine reuptake inhibitor with greater than 99% purity.

  • Molecular Weight: 313.87 g/mol
  • Purity: >99%
  • Form: Solid
  • Activity: Serotonin-norepinephrine reuptake inhibitor (antidepressant)
  • In vitro properties: Ki values of 82 nM for SERT and 2480 nM for NET; displays 30-fold higher affinity for SERT than NET

The product is subject to the following: Abcam Restricted Use Statement

Specifikationer

Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Target Molecule Serotonin Transporter, Norepinephrine Transporter
Formulation 100% Venlafaxine hydrochloride
Quantity 10 mg
For Use With (Application) Antidepressant
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.