Learn More
VEGF 121 Antibody Pair (HRP), Novus Biologicals™
Product Code. 18285296 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 Plates
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18285296 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18285296 Supplier Novus Biologicals Supplier No. NBP2788435PLATES
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Rabbit Monoclonal Antibody
VEGF 121 Monoclonal specifically detects VEGF 121 in Human samples. It is validated for ELISA/Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Antigen VEGF 121
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Rabbit
Quantity 5 Plates
Research Discipline Angiogenesis, Biologically Active Proteins
Gene ID (Entrez) 7422
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human VEGF 121.
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.