Learn More
VEGF 121 Antibody Pair (HRP), Novus Biologicals™
Product Code. 18277456 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18277456 15 Plates Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18277456 Supplier Novus Biologicals Supplier No. NBP27884315PLATES
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Rabbit Monoclonal Antibody
VEGF 121 Monoclonal specifically detects VEGF 121 in Human samples. It is validated for ELISA/Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Antigen VEGF 121
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Rabbit
Quantity 15 Plates
Research Discipline Angiogenesis, Biologically Active Proteins
Gene ID (Entrez) 7422
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human VEGF 121.
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.