Learn More
VDAC1/Porin Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
129.00€ - 399.00€
Specifications
| Antigen | VDAC1/Porin |
|---|---|
| Clone | 2F4 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236072
|
Proteintech
81538-1-RR-100UL |
100 μL |
401.60€ 399.00€ / 100µL Save 2.60€ 0.65% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233889
|
Proteintech
81538-1-RR-20UL |
20 μL |
149.00€ 129.00€ / 20µL Save 20.00€ 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
The VDAC1/Porin antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human VDAC1/Porin. This antibody recognizes human, mouse, rat antigen. The VDAC1/Porin antibody has been validated for the following applications: WB, IHC, IF-P, FC (Intra), ELISA analysis.Tekniske data
| VDAC1/Porin | |
| 500 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 7416 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2F4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MGC111064; PORIN; PORIN-31-HL | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel