Learn More
VCAM-1 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30958-1-AP-150UL
 View more versions of this product
Product Code. 18098705
  • 410.00€ / 150µL
Description
The VCAM-1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse VCAM-1. This antibody recognizes mouse, rat antigen. The VCAM-1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
VCAM-1
600 μg/mL
Rabbit
Primary
Mouse, Rat
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
CD106, Vcam 1, Vcam1
150 μL
22329
Store at -20°C. Stable for one year after shipment.