missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASA Antibody (2F9H5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51586-0.025ml
This item is not returnable.
View return policy
Description
VASA Monoclonal specifically detects VASA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| VASA | |
| Monoclonal | |
| Unconjugated | |
| Q9NQI0 | |
| DDX4 | |
| Purified recombinant fragment of human DDX4 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 54514 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 2F9H5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 4, DEAD box protein 4, EC 3.6.1, EC 3.6.4.13, MGC111074, probable ATP-dependent RNA helicase DDX4, Vasa homolog, VASADEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4 | |
| Mouse | |
| 76 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction