Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Product Code. 15717377 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2.0 mL
0.5 to 5.0 mL
1.0 to 10 mL
10 to 100 mL
2.5 to 25 mL
5.0 to 50 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15717377 2.5 to 25 mL Each
15787367 0.2 to 2.0 mL Each
15797367 0.5 to 5.0 mL Each
15707377 1.0 to 10 mL Each
15781587 5.0 to 50 mL Each
15727377 10 to 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15717377 Supplier Eppendorf™ Supplier No. 4967000049
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
Bulk Quote Request
Ideal for dispensing aliquots of liquid from large supply bottles.
  • Recirculation valve
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
TRUSTED_SUSTAINABILITY
Specifications
Product Type Analog Bottletop Dispenser with Adapters
Channels 1
Volume (Metric) 2.5 to 25 mL
Includes Recirculation valve, adapters GL 32, GL 38, S 40, and telescopic intake tube (170 – 330 mm)