Learn More
Eppendorf™ Varispenser™ 2, Bottle-Top Dispenser
Product Code. 15737357 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 25 mL
5.0 to 50 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15737357 2.5 to 25 mL Each
15707357 0.2 to 2 mL Each
15717357 0.5 to 5 mL Each
15727357 1 to 10 mL Each
15747357 5.0 to 50 mL Each
15757357 10 to 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15737357 Supplier Eppendorf™ Supplier No. 4966000045
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
Bulk Quote Request
Varispenser™ 2, Bottle-Top Dispenser is ideal for dispensing aliquots of liquid from large supply bottles.
Varispenser™ 2 is ideal for dispensing aliquots of liquid from large supply bottles.
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
TRUSTED_SUSTAINABILITY
Specifications
Product Type Analog Bottletop Dispenser with Adapters
Channels 1
Volume (Metric) 2.5 to 25 mL
Includes Adapters GL 32, GL 38, S 40, and telescopic intake tube (170 to 330 mm)